Assay Design



Gene Transcripts

Gene Symbol
Oprl1
Entrez Gene ID
29256
Gene Name
opiate receptor-like 1
Gene Aliases
KOR-3, KOR3, LC132, MOR-C, OFQR, ORL1, Oprl, XOR1
Chromosome Location
Chr.3: 177225366 - 177231663 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006235739.2 XP_006235801.1 4 - 5 589 97
GenBank mRNA AF115267.1 - 2 - 3 286 97

View Full Product Details