Assay Design



Gene Transcripts

Gene Symbol
Jmjd6
Entrez Gene ID
360665
Gene Name
jumonji domain containing 6
Gene Aliases
Ptdsr
Chromosome Location
Chr.10: 105781752 - 105787803 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006247806.1 XP_006247868.1 4 - 5 1027 107

View Full Product Details