Assay Design



Gene Transcripts

Gene Symbol
Lsm12
Entrez Gene ID
287731
Gene Name
LSM12 homolog (S. cerevisiae)
Gene Aliases
RGD1309441
Chromosome Location
Chr.10: 90105562 - 90127600 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105843.1 NP_001099313.1 2 - 3 311 71

View Full Product Details