Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108982.2 | NP_001102452.2 | 3 - 4 | 520 | 123 |
XM_006240104.2 | XP_006240166.1 | 3 - 4 | 607 | 123 | |
GenBank mRNA | FQ228820.1 | - | 3 - 4 | 523 | 123 |
FQ228857.1 | - | 3 - 4 | 527 | 123 |