Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033707.1 | NP_001028879.1 | 9 - 10 | 848 | 91 |
XM_006234529.2 | XP_006234591.1 | 10 - 11 | 1022 | 91 | |
XM_006234530.2 | XP_006234592.1 | 10 - 11 | 994 | 91 | |
XM_006234531.2 | XP_006234593.1 | 9 - 10 | 979 | 91 | |
GenBank mRNA | BC101917.1 | - | 9 - 10 | 848 | 91 |
FQ218185.1 | - | 9 - 10 | 920 | 91 | |
FQ228393.1 | - | 8 - 9 | 1881 | 91 |