Assay Design



Gene Transcripts

Gene Symbol
Immp1l
Entrez Gene ID
691145
Gene Name
IMP1 inner mitochondrial membrane peptidase-like (S. cerevisiae)
Gene Aliases
-
Chromosome Location
Chr.3: 95955125 - 96024316 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_001076990.5 XP_001076990.3 3 - 4 354 98
XM_006234669.2 XP_006234731.1 3 - 4 354 98

View Full Product Details