Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_080584.4 | NP_542151.1 | 3 - 4 | 671 | 94 |
XM_008759855.1 | XP_008758077.1 | 4 - 5 | 494 | 94 | |
XM_008759856.1 | XP_008758078.1 | 4 - 5 | 612 | 94 | |
GenBank mRNA | BC107906.1 | - | 3 - 4 | 531 | 94 |
BC128715.1 | - | 3 - 4 | 600 | 94 | |
M73808.1 | - | 3 - 4 | 671 | 94 |