Assay Design



Gene Transcripts

Gene Symbol
Cir1
Entrez Gene ID
362149
Gene Name
corepressor interacting with RBPJ, 1
Gene Aliases
1700023b02rik, Cir, RGD1309199
Chromosome Location
Chr.3: 59994547 - 60023843 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007799.1 NP_001007800.1 7 - 8 486 83
GenBank mRNA BC085869.1 - 7 - 8 556 83

View Full Product Details