Assay Design



Gene Transcripts

Gene Symbol
Slco4c1
Entrez Gene ID
432363
Gene Name
solute carrier organic anion transporter family, member 4C1
Gene Aliases
Oatp-H, Oatp-M1, Oatp4c1, Slc21A20
Chromosome Location
Chr.9: 104301935 - 104350262 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002024.1 NP_001002024.1 5 - 6 1004 91
GenBank mRNA AF454761.1 - 5 - 6 1004 91

View Full Product Details