Assay Design



Gene Transcripts

Gene Symbol
Ankrd54
Entrez Gene ID
362957
Gene Name
ankyrin repeat domain 54
Gene Aliases
RGD1309552
Chromosome Location
Chr.7: 120282423 - 120295646 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025285.1 NP_001020456.1 7 - 8 884 57
XM_006242012.2 XP_006242074.1 7 - 8 957 57
GenBank mRNA BC093616.1 - 7 - 8 884 57

View Full Product Details