Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006242421.2 | XP_006242483.1 | 11 - 12 | 1032 | 92 |
XM_006242422.2 | XP_006242484.1 | 11 - 12 | 1020 | 92 | |
XM_006242426.2 | XP_006242488.1 | 10 - 11 | 939 | 92 | |
XM_006242427.2 | XP_006242489.1 | 10 - 11 | 927 | 92 |