Assay Design



Gene Transcripts

Gene Symbol
Lhx8
Entrez Gene ID
365963
Gene Name
LIM homeobox 8
Gene Aliases
Lhx7
Chromosome Location
Chr.2: 260572827 - 260598744 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012219.1 NP_001012219.1 1 - 2 143 126
XM_006233528.2 XP_006233590.1 2 - 3 409 126
GenBank mRNA BC090011.1 - 1 - 2 143 126

View Full Product Details