Assay Design



Gene Transcripts

Gene Symbol
Meis2
Entrez Gene ID
311311
Gene Name
Meis homeobox 2
Gene Aliases
-
Chromosome Location
Chr.3: 107560172 - 107762704 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107758.1 NP_001101228.1 6 - 7 875 57
XM_006234742.2 XP_006234804.1 6 - 7 1073 57
XM_006234743.2 XP_006234805.1 7 - 8 1085 57
XM_006234744.2 XP_006234806.1 7 - 8 1119 57
XM_006234745.2 XP_006234807.1 7 - 8 1051 57
XM_006234746.2 XP_006234808.1 6 - 7 1073 57
XM_006234747.2 XP_006234809.1 6 - 7 862 57
XM_006234748.2 XP_006234810.1 6 - 7 1196 57
XM_006234749.2 XP_006234811.1 6 - 7 862 57
XM_006234750.2 XP_006234812.1 6 - 7 1073 57
XM_006234751.2 XP_006234813.1 6 - 7 1073 57
XM_006234752.2 XP_006234814.1 6 - 7 862 57
XM_006234753.2 XP_006234815.1 6 - 7 1196 57
XM_006234754.2 XP_006234816.1 6 - 7 862 57
XM_006234755.2 XP_006234817.1 6 - 7 1196 57
XM_008762094.1 XP_008760316.1 3 - 4 367 57

View Full Product Details