Assay Design



Gene Transcripts

Gene Symbol
Camk1d
Entrez Gene ID
307124
Gene Name
calcium/calmodulin-dependent protein kinase ID
Gene Aliases
RGD1560691
Chromosome Location
Chr.17: 76532611 - 76938956 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107365.1 NP_001100835.1 11 - 12 1194 83

View Full Product Details