Assay Design



Gene Transcripts

Gene Symbol
Cat
Entrez Gene ID
24248
Gene Name
catalase
Gene Aliases
CS1, Cas1, Cat01, Catl, Cs-1
Chromosome Location
Chr.3: 93379872 - 93412058 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012520.2 NP_036652.1 9 - 10 1324 67
GenBank mRNA BC081853.1 - 9 - 10 1221 67
M11670.1 - 9 - 10 1275 67

View Full Product Details