Assay Design



Gene Transcripts

Gene Symbol
Prdm2
Entrez Gene ID
313678
Gene Name
PR domain containing 2, with ZNF domain
Gene Aliases
Riz
Chromosome Location
Chr.5: 161769587 - 161879134 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001077648.1 NP_001071116.1 1 - 2 92 131
GenBank mRNA U17837.1 - 1 - 2 92 131

View Full Product Details