Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077648.1 | NP_001071116.1 | 5 - 6 | 539 | 94 |
XM_006239335.2 | XP_006239397.1 | 6 - 7 | 622 | 94 | |
XM_006239337.2 | XP_006239399.1 | 2 - 3 | 209 | 94 | |
XM_008764255.1 | XP_008762477.1 | 5 - 6 | 4173 | 94 | |
GenBank mRNA | U17837.1 | - | 5 - 6 | 539 | 94 |