Assay Design



Gene Transcripts

Gene Symbol
Igf2bp2
Entrez Gene ID
303824
Gene Name
insulin-like growth factor 2 mRNA binding protein 2
Gene Aliases
RGD1305614
Chromosome Location
Chr.11: 82466071 - 82621105 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270598.1 NP_001257527.1 15 - 16 1762 66
NM_001270599.1 NP_001257528.1 15 - 16 1762 66
GenBank mRNA FQ222567.1 - 15 - 16 1530 66

View Full Product Details