Assay Design



Gene Transcripts

Gene Symbol
Xkrx
Entrez Gene ID
497101
Gene Name
XK, Kell blood group complex subunit-related, X-linked
Gene Aliases
XRG2
Chromosome Location
Chr.X: 104973083 - 104984261 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012230.1 NP_001012230.1 1 - 2 456 76
GenBank mRNA AY534257.1 - 1 - 2 456 76

View Full Product Details