Assay Design



Gene Transcripts

Gene Symbol
Syt5
Entrez Gene ID
54309
Gene Name
synaptotagmin V
Gene Aliases
SytIX
Chromosome Location
Chr.1: 72860218 - 72867934 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019350.2 NP_062223.1 6 - 7 1026 114
GenBank mRNA BC092198.1 - 6 - 7 1026 114
U26402.1 - 6 - 7 829 114
X84884.1 - 5 - 6 779 114

View Full Product Details