Assay Design



Gene Transcripts

Gene Symbol
Prkag3
Entrez Gene ID
301518
Gene Name
protein kinase, AMP-activated, gamma 3 non-catalytic subunit
Gene Aliases
Ampkg3
Chromosome Location
Chr.9: 81999368 - 82008663 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106921.1 NP_001100391.1 8 - 9 987 147
XM_008767222.1 XP_008765444.1 8 - 9 1018 135

View Full Product Details