Assay Design



Gene Transcripts

Gene Symbol
Wnt6
Entrez Gene ID
316526
Gene Name
wingless-type MMTV integration site family, member 6
Gene Aliases
-
Chromosome Location
Chr.9: 82033543 - 82047172 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108226.1 NP_001101696.1 2 - 3 542 73

View Full Product Details