Assay Design



Gene Transcripts

Gene Symbol
Impact
Entrez Gene ID
497198
Gene Name
impact RWD domain protein
Gene Aliases
-
Chromosome Location
Chr.18: 4325872 - 4346734 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012235.1 NP_001012235.1 2 - 3 245 98
GenBank mRNA AY574213.1 - 2 - 3 245 98

View Full Product Details