Assay Design



Gene Transcripts

Gene Symbol
Mreg
Entrez Gene ID
501162
Gene Name
melanoregulin
Gene Aliases
RGD1562333
Chromosome Location
Chr.9: 79490058 - 79545024 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001192002.1 NP_001178931.1 4 - 5 509 98
XM_006245265.2 XP_006245327.1 5 - 6 657 98

View Full Product Details