Assay Design



Gene Transcripts

Gene Symbol
Crygf
Entrez Gene ID
689947
Gene Name
crystallin, gamma F
Gene Aliases
Cryg6, Len
Chromosome Location
Chr.9: 72933304 - 72935468 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001109557.1 NP_001103027.1 1 - 2 105 61

View Full Product Details