Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108906.1 | NP_001102376.1 | 3 - 4 | 524 | 92 |
XM_006228118.1 | XP_006228180.1 | 3 - 4 | 914 | 92 | |
GenBank mRNA | BC167093.1 | - | 3 - 4 | 522 | 92 |
FQ222002.1 | - | 3 - 4 | 490 | 92 | |
FQ227339.1 | - | 3 - 4 | 540 | 92 | |
FQ230074.1 | - | 3 - 4 | 525 | 92 | |
FQ230732.1 | - | 3 - 4 | 540 | 92 | |
FQ231147.1 | - | 3 - 4 | 519 | 92 | |
FQ233974.1 | - | 3 - 4 | 435 | 92 |