Assay Design



Gene Transcripts

Gene Symbol
Ube2m
Entrez Gene ID
361509
Gene Name
ubiquitin-conjugating enzyme E2M
Gene Aliases
-
Chromosome Location
Chr.1: 65537640 - 65541177 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108471.1 NP_001101941.1 2 - 3 775 68
GenBank mRNA FQ209516.1 - 3 - 4 368 68

View Full Product Details