Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006257680.2 | XP_006257742.1 | 9 - 10 | 3277 | 75 |
XM_006257681.2 | XP_006257743.1 | 10 - 11 | 2385 | 75 | |
XM_006257682.2 | XP_006257744.1 | 9 - 10 | 3274 | 75 | |
XM_006257683.2 | XP_006257745.1 | 9 - 10 | 3409 | 75 | |
XM_008773648.1 | XP_008771870.1 | 7 - 8 | 2956 | 75 |