Assay Design
Gene Transcripts
- Gene Symbol
- Chmp2b
- Entrez Gene ID
-
363720
- Gene Name
- charged multivesicular body protein 2B
- Gene Aliases
-
RGD1306781
- Chromosome Location
-
Chr.11: 2665264 - 2692254 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_343995.7 |
XP_343996.5 |
2 - 3 |
411 |
85 |
GenBank mRNA |
FQ212834.1 |
- |
2 - 3 |
376 |
85 |
View Full Product Details