Assay Design



Gene Transcripts

Gene Symbol
Gbe1
Entrez Gene ID
288333
Gene Name
glucan (1,4-alpha-), branching enzyme 1
Gene Aliases
-
Chromosome Location
Chr.11: 7210169 - 7485895 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001100502.1 NP_001093972.1 5 - 6 824 121
GenBank mRNA BC090037.1 - 2 - 3 199 121

View Full Product Details