Assay Design



Gene Transcripts

Gene Symbol
Hars
Entrez Gene ID
307492
Gene Name
histidyl-tRNA synthetase
Gene Aliases
Dnd1
Chromosome Location
Chr.18: 29611545 - 29629087 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001025414.1 NP_001020585.1 2 - 3 199 78
GenBank mRNA BC097969.1 - 2 - 3 199 78

View Full Product Details