Assay Design



Gene Transcripts

Gene Symbol
Ttc33
Entrez Gene ID
294774
Gene Name
tetratricopeptide repeat domain 33
Gene Aliases
RGD1564042
Chromosome Location
Chr.2: 54897363 - 54944241 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006231994.1 XP_006232056.1 1 - 2 58 66

View Full Product Details