Assay Design



Gene Transcripts

Gene Symbol
Sh2d4a
Entrez Gene ID
306376
Gene Name
SH2 domain containing 4A
Gene Aliases
-
Chromosome Location
Chr.16: 23084247 - 23156825 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012048.1 NP_001012048.1 3 - 4 564 97
XM_006253032.2 XP_006253094.1 3 - 4 577 97
GenBank mRNA BC079100.1 - 3 - 4 564 97

View Full Product Details