Assay Design



Gene Transcripts

Gene Symbol
Tma16
Entrez Gene ID
290686
Gene Name
translation machinery associated 16 homolog (S. cerevisiae)
Gene Aliases
RGD1305222
Chromosome Location
Chr.16: 24978431 - 24997286 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169102.1 NP_001162573.1 3 - 4 202 129
XM_006253023.2 XP_006253085.1 3 - 4 298 129
GenBank mRNA CF109688.1 - 3 - 4 237 129

View Full Product Details