Assay Design



Gene Transcripts

Gene Symbol
Rab6b
Entrez Gene ID
363123
Gene Name
RAB6B, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.8: 111600372 - 111668817 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108775.1 NP_001102245.1 3 - 4 214 112
XM_006243669.2 XP_006243731.1 4 - 5 518 112

View Full Product Details