Assay Design



Gene Transcripts

Gene Symbol
Osbpl10
Entrez Gene ID
316039
Gene Name
oxysterol binding protein-like 10
Gene Aliases
RGD1561652
Chromosome Location
Chr.8: 123029728 - 123293898 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006244025.2 XP_006244087.1 8 - 9 1761 98
XM_008766657.1 XP_008764879.1 8 - 9 1818 98
XM_243980.7 XP_243980.4 8 - 9 2913 98

View Full Product Details