Assay Design



Gene Transcripts

Gene Symbol
Tfdp2
Entrez Gene ID
300947
Gene Name
transcription factor Dp-2 (E2F dimerization partner 2)
Gene Aliases
Tcfdp2
Chromosome Location
Chr.8: 104040796 - 104181228 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001106847.2 NP_001100317.2 8 - 9 867 87
XM_006243556.2 XP_006243618.1 - 1171 87
XM_006243558.2 XP_006243620.1 8 - 9 2270 87
XM_006243561.2 XP_006243623.1 7 - 8 2230 87
XM_006243563.1 XP_006243625.1 10 - 11 1279 87
XM_006243566.2 XP_006243628.1 - 1472 87
XM_006243567.2 XP_006243629.1 9 - 10 2594 87
XM_006243568.2 XP_006243630.1 10 - 11 2691 87
XM_006243569.2 XP_006243631.1 9 - 10 1405 87
XM_006243571.2 XP_006243633.1 9 - 10 1358 87
XM_006243572.2 XP_006243634.1 7 - 8 2226 87
XM_008766479.1 XP_008764701.1 9 - 10 849 87
XM_008766480.1 XP_008764702.1 9 - 10 845 87
XM_008766481.1 XP_008764703.1 9 - 10 849 87
XM_008766482.1 XP_008764704.1 13 - 14 1386 87
XM_008766483.1 XP_008764705.1 9 - 10 1172 87
XM_008766484.1 XP_008764706.1 13 - 14 1240 87
XM_008766485.1 XP_008764707.1 12 - 13 1144 87
XM_008766487.1 XP_008764709.1 10 - 11 975 87
XM_008766488.1 XP_008764710.1 7 - 8 685 87
GenBank mRNA BC168961.1 - 8 - 9 867 87

View Full Product Details