Assay Design



Gene Transcripts

Gene Symbol
Fam83b
Entrez Gene ID
315829
Gene Name
family with sequence similarity 83, member B
Gene Aliases
RGD1311988
Chromosome Location
Chr.8: 83616148 - 83693484 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108169.1 NP_001101639.1 2 - 3 558 134

View Full Product Details