Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001108772.1 | NP_001102242.1 | 2 - 3 | 321 | 63 |
XM_006243482.2 | XP_006243544.1 | 2 - 3 | 333 | 63 | |
XM_008766451.1 | XP_008764673.1 | 2 - 3 | 349 | 63 | |
GenBank mRNA | BC166838.1 | - | 2 - 3 | 243 | 63 |