Assay Design
Gene Transcripts
- Gene Symbol
- Rbm15b
- Entrez Gene ID
-
315988
- Gene Name
- RNA binding motif protein 15B
- Gene Aliases
-
RGD1305937
- Chromosome Location
-
Chr.8: 115618186 - 115622846 on Build Rnor_6.0
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
XM_236613.9 |
XP_236613.6 |
2 - 2 |
4249 |
121 |
View Full Product Details