Assay Design



Gene Transcripts

Gene Symbol
Ephb3
Entrez Gene ID
287989
Gene Name
Eph receptor B3
Gene Aliases
-
Chromosome Location
Chr.11: 83527962 - 83546802 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001105868.1 NP_001099338.1 4 - 5 1402 88
XM_006248561.2 XP_006248623.1 4 - 5 1530 88

View Full Product Details