Assay Design



Gene Transcripts

Gene Symbol
Fetub
Entrez Gene ID
83928
Gene Name
fetuin B
Gene Aliases
Fet, Pp63
Chromosome Location
Chr.11: 81648890 - 81661915 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053348.1 NP_445800.1 5 - 6 809 83
XM_006248549.1 XP_006248611.1 6 - 7 763 83
XM_006248550.1 XP_006248612.1 6 - 7 1126 83
GenBank mRNA AJ242926.1 - 5 - 6 809 83
BC070508.1 - 6 - 7 748 83
FQ209610.1 - 5 - 6 796 83
FQ209832.1 - 5 - 6 761 83
FQ210335.1 - 5 - 6 83
FQ211050.1 - 5 - 6 760 83
FQ218930.1 - 5 - 6 793 83
FQ219048.1 - 5 - 6 791 83
FQ219266.1 - 5 - 6 760 83
FQ219352.1 - 5 - 6 760 83
FQ219426.1 - 3 - 4 362 83
FQ219454.1 - 5 - 6 760 83
FQ219500.1 - 5 - 6 763 83
FQ219513.1 - 5 - 6 832 83

View Full Product Details