Assay Design



Gene Transcripts

Gene Symbol
Fetub
Entrez Gene ID
83928
Gene Name
fetuin B
Gene Aliases
Fet, Pp63
Chromosome Location
Chr.11: 81648890 - 81661915 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053348.1 NP_445800.1 6 - 7 908 70
XM_006248549.1 XP_006248611.1 7 - 8 862 70
XM_006248550.1 XP_006248612.1 7 - 8 1225 70
GenBank mRNA AJ242926.1 - 6 - 7 908 70
BC070508.1 - 7 - 8 847 70
FQ209398.1 - 6 - 7 920 70
FQ209518.1 - 6 - 7 859 70
FQ209610.1 - 6 - 7 895 70
FQ209832.1 - 6 - 7 860 70
FQ210335.1 - 6 - 7 860 70
FQ211050.1 - 6 - 7 859 70
FQ218884.1 - 7 - 8 70
FQ218893.1 - 5 - 6 760 70
FQ218905.1 - 6 - 7 859 70
FQ218930.1 - 6 - 7 892 70
FQ218949.1 - 6 - 7 709 70
FQ219048.1 - 6 - 7 890 70
FQ219152.1 - 6 - 7 896 70
FQ219170.1 - 6 - 7 859 70
FQ219190.1 - 6 - 7 895 70
FQ219266.1 - 6 - 7 859 70
FQ219352.1 - 6 - 7 859 70
FQ219426.1 - 4 - 5 461 70
FQ219454.1 - 6 - 7 859 70
FQ219459.1 - 6 - 7 858 70
FQ219500.1 - 6 - 7 862 70

View Full Product Details