Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001013864.1 | NP_001013886.1 | 5 - 6 | 539 | 62 |
XM_006248505.2 | XP_006248567.1 | 8 - 9 | 976 | 62 | |
GenBank mRNA | BC083627.1 | - | 5 - 6 | 539 | 62 |
BC083890.1 | - | 4 - 5 | 477 | 62 |