Assay Design



Gene Transcripts

Gene Symbol
Slc7a4
Entrez Gene ID
303787
Gene Name
solute carrier family 7, member 4
Gene Aliases
-
Chromosome Location
Chr.11: 87445585 - 87450006 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_006248693.2 XP_006248755.1 2 - 3 386 73
XM_006248694.2 XP_006248756.1 2 - 3 206 73

View Full Product Details