Assay Design



Gene Transcripts

Gene Symbol
Dvl3
Entrez Gene ID
303811
Gene Name
dishevelled segment polarity protein 3
Gene Aliases
-
Chromosome Location
Chr.11: 84051171 - 84068318 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001107081.2 NP_001100551.1 2 - 3 82 69
XM_006248564.2 XP_006248626.1 2 - 3 313 69
XM_006248565.2 XP_006248627.1 2 - 3 241 69
XM_006248566.2 XP_006248628.1 2 - 3 241 69

View Full Product Details