Assay Design



Gene Transcripts

Gene Symbol
Dgcr14
Entrez Gene ID
360741
Gene Name
DiGeorge syndrome critical region gene 14
Gene Aliases
-
Chromosome Location
Chr.11: 87224373 - 87234329 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012472.1 NP_001012490.1 8 - 9 1050 104
XM_006248635.2 XP_006248697.1 8 - 9 1095 104
XM_006248636.2 XP_006248698.1 7 - 8 990 104
GenBank mRNA BC089898.1 - 8 - 9 1050 104

View Full Product Details