Assay Design



Gene Transcripts

Gene Symbol
Parl
Entrez Gene ID
287979
Gene Name
presenilin associated, rhomboid-like
Gene Aliases
Psarl
Chromosome Location
Chr.11: 84517368 - 84544463 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035249.1 NP_001030326.1 2 - 3 363 75
GenBank mRNA BC105907.1 - 2 - 3 363 75
BC126098.1 - 2 - 3 339 75
BC158564.1 - 2 - 3 347 75
FQ214115.1 - 2 - 3 358 75

View Full Product Details