Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035249.1 | NP_001030326.1 | 4 - 5 | 551 | 69 |
GenBank mRNA | BC088226.1 | - | 4 - 5 | 469 | 69 |
BC105907.1 | - | 4 - 5 | 551 | 69 | |
BC158564.1 | - | 4 - 5 | 535 | 69 | |
FQ214115.1 | - | 4 - 5 | 546 | 69 |