Assay Design



Gene Transcripts

Gene Symbol
Tbx1
Entrez Gene ID
360737
Gene Name
T-box 1
Gene Aliases
-
Chromosome Location
Chr.11: 86552022 - 86561647 on Build Rnor_6.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001108322.1 NP_001101792.1 2 - 3 165 67
XM_006248619.2 XP_006248681.1 1 - 2 947 67

View Full Product Details